(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate 97%

Supplier: Ambeed
Warning

Synonyms: (2,2-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate

A686505-100MG A686505-1G A686505-250MG
76651-034EA 43 USD
76651-034 76651-036 76651-038
(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate 97%
(2,2'-Bipyridine)bis[2-(2,4-difluorophenyl)pyridine]iridium(III) Hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₂H₂₀F₁₀IrN₄P
MW: 873.71 g/mol
MDL Number: MFCD29472513
CAS Number: 864163-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR