2-Fluoro-4-(propan-2-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A686094-1G A686094-250MG
76651-030EA 209 USD
76651-030 76651-032
2-Fluoro-4-(propan-2-yl)benzoic acid 95%
2-Fluoro-4-(propan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.1916 g/mol
MDL Number: MFCD19598690
CAS Number: 1341179-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR