SM 16 97%

Supplier: Ambeed
Warning

A685977-100MG A685977-10MG A685977-1MG A685977-25MG A685977-50MG A685977-5MG
76651-018EA 243 USD
76651-018 76651-020 76651-022 76651-024 76651-026 76651-028
SM 16 97%
SM 16

Precaución: for Research Use Only

Formula: C₂₅H₂₆N₄O₃
MDL Number: MFCD31657399
CAS Number: 614749-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR