tert-butyl N-[4-amino-4-(cyanomethyl)cyclohexyl]carbamate 98%

Supplier: Ambeed
Warning

A684934-1G A684934-25MG A684934-50MG
76651-010EA 474 USD
76651-010 76651-012 76651-014
tert-butyl N-[4-amino-4-(cyanomethyl)cyclohexyl]carbamate 98%
tert-butyl N-[4-amino-4-(cyanomethyl)cyclohexyl]carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₃N₃O₂
CAS Number: 1656989-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR