(S)-2-Amino-3-(4-(trifluoromethyl)phenyl)propan-1-ol 95%

Supplier: Ambeed
Warning

A681941-100MG A681941-1G A681941-250MG
76650-994EA 101.85 USD
76650-994 76650-996 76650-998
(S)-2-Amino-3-(4-(trifluoromethyl)phenyl)propan-1-ol 95%
(S)-2-Amino-3-(4-(trifluoromethyl)phenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₃NO
MW: 219.2036 g/mol
MDL Number: MFCD17171136
CAS Number: 153181-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR