tert-Butyl (17-bromo-3,6,9,12,15-pentaoxaheptadecyl)carbamate 95%

Supplier: Ambeed
Warning

A681582-100MG A681582-1G A681582-250MG A681582-50MG
76650-988EA 40 USD
76650-988 76650-990 76650-992 77622-454
tert-Butyl (17-bromo-3,6,9,12,15-pentaoxaheptadecyl)carbamate 95%
tert-Butyl (17-bromo-3,6,9,12,15-pentaoxaheptadecyl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₃₄BrNO₇
MDL Number: MFCD31580050
CAS Number: 1392499-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR