ML239 97%

Supplier: Ambeed
Danger

Synonyms: (E)-N'-((1H-Pyrrol-2-yl)methylene)-2-(2,4,6-trichlorophenoxy)acetohydrazide, CID-49843203, (2,4,6-Trichlorophenoxy)-acetic acid (1H-pyrrol-2-ylmethylene)-hydrazide, 2-(2,4,6-Trichlorophenoxy)acetic acid (2E)-2-(1H-pyrrol-2-ylmethylene)hydrazide, N'-[(Z)-pyrrol-2-ylidenemethyl]-2-(2,4,6-trichlorophenoxy)acetohydrazide

A681365-100MG A681365-10MG A681365-1MG A681365-25MG A681365-50MG A681365-5MG
76650-970EA 81.69 USD
76650-970 76650-972 76650-974 76650-976 76650-978 76650-980
ML239 97%
ML239

Precaución: for Research Use Only

Formula: C₁₃H10Cl₃N₃O₂
MW: 346.6 g/mol
MDL Number: MFCD23098913
CAS Number: 1378872-36-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR