5-(3-Bromophenyl)-7-(6-morpholinopyridin-3-yl)pyrido[2,3-d]pyrimidin-4-amine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: ABT-702 2HCl

A680393-100MG A680393-10MG A680393-1MG A680393-50MG A680393-5MG
76650-960EA 264.17 USD
76650-960 76650-962 76650-964 76650-966 76650-968
5-(3-Bromophenyl)-7-(6-morpholinopyridin-3-yl)pyrido[2,3-d]pyrimidin-4-amine dihydrochloride 95%
5-(3-Bromophenyl)-7-(6-morpholinopyridin-3-yl)pyrido[2,3-d]pyrimidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₁BrCl₂N₆O
MW: 536.2517 g/mol
MDL Number: MFCD03452809
CAS Number: 1188890-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR