N-Boc-(R)-2-(methylamino)butyric acid 95%

Supplier: Ambeed
Warning

A679572-100MG A679572-1G A679572-250MG
76650-952EA 6 USD
76650-952 76650-954 76650-956
N-Boc-(R)-2-(methylamino)butyric acid 95%
N-Boc-(R)-2-(methylamino)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD22380682
CAS Number: 287210-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR