3-Fluoro-2-methylbenzeneboronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3,2-Dioxaborolane, 2-(3-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-, 2-(3-Fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-2-methylphenylboronic acid pinacol ester
A679388-100MG
A679388-1G
A679388-250MG
A679388-5G
76650-944EA
37.14
USD
76650-944
76650-946
76650-948
76650-950
3-Fluoro-2-methylbenzeneboronic acid pinacol ester 95%
3-Fluoro-2-methylbenzeneboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BFO₂ MW: 236.09 g/mol |
MDL Number:
MFCD18729892 CAS Number: 1417301-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...