1-Methyl-2-(2-pyridyl)pyridinium iodide 98% (by HPLC, titration analysis)

Supplier: Ambeed
Warning

A679184-100MG A679184-1G A679184-5G A679184-250MG
76650-938EA 10 USD
76650-938 76650-940 76650-942 77783-641
1-Methyl-2-(2-pyridyl)pyridinium iodide 98% (by HPLC, titration analysis)
1-Methyl-2-(2-pyridyl)pyridinium iodide

Precaución: for Research Use Only

Formula: C₁₁H₁₁IN₂
MDL Number: MFCD23700876
CAS Number: 77972-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR