2,6-Dibromo-N,N-dimethylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A678220-250MG A678220-5G A678220-1G
76650-934EA 31 USD
76650-934 76650-936 77749-395
2,6-Dibromo-N,N-dimethylpyridin-4-amine 95%
2,6-Dibromo-N,N-dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₈Br₂N₂
MDL Number: MFCD28672017
CAS Number: 192447-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR