2-(2-((tert-Butoxycarbonyl)(methyl)amino)-N-methylacetamido)acetic acid 95%

Supplier: Ambeed
Warning

A677202-250MG A677202-1G
76650-922EA 55.17 USD
76650-922 77622-446
2-(2-((tert-Butoxycarbonyl)(methyl)amino)-N-methylacetamido)acetic acid 95%
2-(2-((tert-Butoxycarbonyl)(methyl)amino)-N-methylacetamido)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₅
MW: 260.2869 g/mol
MDL Number: MFCD18910599
CAS Number: 56612-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR