4-Fluoro-2-iodopyridine 97%

Supplier: Ambeed
Warning

A675427-1G A675427-250MG
76650-900EA 403.15 USD
76650-900 76650-902
4-Fluoro-2-iodopyridine 97%
4-Fluoro-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃FIN
MW: 222.9869 g/mol
MDL Number: MFCD31728216
CAS Number: 2385268-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR