6-(2-(Methylsulfonyl)pyrimidin-5-yl)-N-(prop-2-yn-1-yl)hex-5-ynamide 99%

Supplier: Ambeed
Warning

A674331-100MG A674331-10G A674331-1G A674331-250MG A674331-5G
76650-876EA 28.65 USD
76650-876 76650-878 76650-880 76650-882 76650-884
6-(2-(Methylsulfonyl)pyrimidin-5-yl)-N-(prop-2-yn-1-yl)hex-5-ynamide 99%
6-(2-(Methylsulfonyl)pyrimidin-5-yl)-N-(prop-2-yn-1-yl)hex-5-ynamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₃S
MW: 305.3522 g/mol
MDL Number: MFCD31715256
CAS Number: 2288710-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR