(R)-3,3-Dimethylbutan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A673926-100MG A673926-250MG A673926-1G
76650-866EA 28.65 USD
76650-866 76650-868 77308-100
(R)-3,3-Dimethylbutan-2-amine hydrochloride 95%
(R)-3,3-Dimethylbutan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆ClN
MW: 137.6509 g/mol
MDL Number: MFCD28501709
CAS Number: 31519-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR