2-Amino-4-(4-methoxyphenyl)-7-(naphthalen-1-yl)-5-oxo-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile 97%

Supplier: Ambeed
Warning

A673822-10MG A673822-1MG A673822-25MG A673822-5MG A673822-50MG
76650-858EA 28 USD
76650-858 76650-860 76650-862 76650-864 77749-389
2-Amino-4-(4-methoxyphenyl)-7-(naphthalen-1-yl)-5-oxo-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile 97%
2-Amino-4-(4-methoxyphenyl)-7-(naphthalen-1-yl)-5-oxo-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile

Precaución: for Research Use Only

Formula: C₂₇H₂₂N₂O₃
MDL Number: MFCD12913298
CAS Number: 1118460-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR