Ethyl 4-bromo-6-methylpicolinate 95%

Supplier: Ambeed
Warning

A673588-100MG A673588-250MG A673588-1G
76650-854EA 61.54 USD
76650-854 76650-856 77573-174
Ethyl 4-bromo-6-methylpicolinate 95%
Ethyl 4-bromo-6-methylpicolinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.0852 g/mol
MDL Number: MFCD20485650
CAS Number: 947179-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR