4-(2-Furyl)-3-buten-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Furan-2-yl)but-3-en-2-one, Furfuralacetone

A672216-100G A672216-1G A672216-25G A672216-5G
76650-822EA 143.23 USD
76650-822 76650-824 76650-826 76650-828
4-(2-Furyl)-3-buten-2-one 95%
4-(2-Furyl)-3-buten-2-one

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00039566
CAS Number: 623-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR