2-(Difluoromethoxy)pyridine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Difluoromethoxy)nicotinaldehyde

A671932-100MG A671932-1G A671932-250MG
76650-816EA 217.49 USD
76650-816 76650-818 76650-820
2-(Difluoromethoxy)pyridine-3-carbaldehyde 95%
2-(Difluoromethoxy)pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
MDL Number: MFCD31621142
CAS Number: 2222104-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR