2-Amino-5-bromo-3-methylphenol 95%

Supplier: Ambeed
Warning

A670457-1G A670457-100MG A670457-5G
76650-800EA 246.13 USD
76650-800 77749-379 77749-380
2-Amino-5-bromo-3-methylphenol 95%
2-Amino-5-bromo-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD22381690
CAS Number: 1194760-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR