5-Methylpyridine-3-sulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 5-Methyl-3-pyridinesulphonyl chloride

A664016-1G A664016-100MG A664016-250MG
76650-732EA 740 USD
76650-732 77598-680 77783-615
5-Methylpyridine-3-sulfonyl chloride 97%
5-Methylpyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD13188992
CAS Number: 166337-57-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR