Methyl 5-bromo-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A663928-1G A663928-250MG A663928-5G A663928-100MG
76650-728EA 100 USD
76650-728 76650-730 77569-262 77749-367
Methyl 5-bromo-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzoate 98%
Methyl 5-bromo-3-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-2-methylbenzoate

Precaución: For research use only.

Formula: C₁₆H₂₂BrNO₃
MDL Number: MFCD28347529
CAS Number: 1403257-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR