7-(((Benzyloxy)carbonyl)amino)heptanoic acid 95%

Supplier: Ambeed
Warning

A662365-1G A662365-250MG A662365-5G
76650-720EA 28.65 USD
76650-720 76650-722 76650-724
7-(((Benzyloxy)carbonyl)amino)heptanoic acid 95%
7-(((Benzyloxy)carbonyl)amino)heptanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.3315 g/mol
MDL Number: MFCD09930580
CAS Number: 23434-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR