7-Bromopyrido[3,2-d]pyrimidin-4-ol 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromopyrido[3,2-d]pyrimidin-4(3H)-one

A661142-100MG A661142-10G A661142-1G A661142-250MG A661142-5G
76650-710EA 65.78 USD
76650-710 76650-712 76650-714 76650-716 76650-718
7-Bromopyrido[3,2-d]pyrimidin-4-ol 95%
7-Bromopyrido[3,2-d]pyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O
MW: 226.04 g/mol
MDL Number: MFCD12755861
CAS Number: 573675-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR