Methyl 2-hydroxy-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A660682-100MG A660682-250MG
76650-706EA 99.73 USD
76650-706 76650-708
Methyl 2-hydroxy-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 95%
Methyl 2-hydroxy-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.1815 g/mol
MDL Number: MFCD02177454
CAS Number: 224313-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR