Polyethylene glycol monomethyl ether, M.W. ∼350

Supplier: Ambeed
Warning

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

A660194-1000G A660194-25G A660194-500G A660194-100G
76650-700EA 42 USD
76650-700 76650-702 76650-704 77783-612
Polyethylene glycol monomethyl ether, M.W. ∼350
Polyethylene glycol monomethyl ether

Precaución: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR