6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,3]dioxolo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A658995-50MG A658995-1G A658995-100MG
76650-698EA 38.2 USD
76650-698 77552-012 77617-756
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,3]dioxolo[4,5-b]pyridine 95%
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,3]dioxolo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BNO₄
MW: 249.07 g/mol
MDL Number: MFCD29038168
CAS Number: 2828447-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR