7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: S 107 HCl

A657879-10MG A657879-50MG A657879-5MG A657879-1MG A657879-100MG
76650-688EA 62.6 USD
76650-688 76650-690 76650-692 77598-672 77749-355
7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride 97%
7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClNOS
MW: 245.77 g/mol
MDL Number: MFCD18086903
CAS Number: 1357476-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR