[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol 95%

Supplier: Ambeed
Warning

A657267-100MG A657267-1G A657267-250MG
76650-682EA 36.08 USD
76650-682 76650-684 76650-686
[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol 95%
[4-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.0900 g/mol
MDL Number: MFCD18729956
CAS Number: 1333264-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR