2-(2-Chloro-5-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A656728-100MG A656728-1G A656728-250MG A656728-5G
76650-664EA 34 USD
76650-664 76650-666 76650-668 76650-670
2-(2-Chloro-5-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(2-Chloro-5-fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₂
MDL Number: MFCD29088164
CAS Number: 2121512-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR