2-Fluoro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A653233-1G
A653233-250MG
A653233-5G
76650-640EA
217
USD
76650-640
76650-642
76650-644
2-Fluoro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
2-Fluoro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BFO₃ |
MDL Number:
MFCD28154565 CAS Number: 2121513-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...