2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A650631-100MG A650631-1G A650631-250MG A650631-5G
76650-624EA 36.08 USD
76650-624 76650-626 76650-628 76650-630
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BCl₂O₃
MW: 302.99 g/mol
MDL Number: MFCD22415026
CAS Number: 2121513-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR