6-Bromo-4-methylnicotinamide 95%

Supplier: Ambeed
Danger

A649331-1G A649331-250MG
76650-620EA 135.8 USD
76650-620 76650-622
6-Bromo-4-methylnicotinamide 95%
6-Bromo-4-methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MW: 215.0473 g/mol
CAS Number: 2385048-34-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR