tert-Butyl (26-amino-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG8-CH2CH2NH2

A647409-100MG A647409-1G A647409-250MG A647409-5G
76650-600EA 44 USD
76650-600 76650-602 76650-604 77622-416
tert-Butyl (26-amino-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%
tert-Butyl (26-amino-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₄₈N₂O₁₀
MW: 512.6346 g/mol
MDL Number: MFCD31656919
CAS Number: 1052207-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR