Diphenyl Phenylphosphonate 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Phenylphosphonic acid Diphenyl ester

A646118-100G A646118-25G A646118-5G
76650-588EA 146.41 USD
76650-588 76650-590 76650-592
Diphenyl Phenylphosphonate 98% (by GC)
Diphenyl Phenylphosphonate

Precaución: for Research Use Only

Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
MDL Number: MFCD00216888
CAS Number: 3049-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR