2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-ethoxy-5-fluorophenylboronic acid pinacol ester

A645900-100MG A645900-1G A645900-250MG
76650-582EA 18 USD
76650-582 76650-584 76650-586
2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₉BClFO₃
MDL Number: MFCD24368960
CAS Number: 1668474-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR