Ethyl 2-bromo-1-methyl-1H-imidazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A645600-1G
A645600-100MG
A645600-250MG
76650-578EA
827.51
USD
76650-578
77749-330
77749-331
Ethyl 2-bromo-1-methyl-1H-imidazole-4-carboxylate 97%
Ethyl 2-bromo-1-methyl-1H-imidazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉BrN₂O₂ MW: 233.06 g/mol |
MDL Number:
MFCD30187875 CAS Number: 1639289-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...