2-((tert-Butoxycarbonyl)amino)-2-(3,5-dichlorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A644218-100MG A644218-1G A644218-250MG
76650-572EA 55.17 USD
76650-572 76650-574 76650-576
2-((tert-Butoxycarbonyl)amino)-2-(3,5-dichlorophenyl)acetic acid 95%
2-((tert-Butoxycarbonyl)amino)-2-(3,5-dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅Cl₂NO₄
MW: 320.1700 g/mol
MDL Number: MFCD02683162
CAS Number: 369403-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR