tert-Butyl 4-(2-aminopyridin-4-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A642847-100MG A642847-1G A642847-250MG A642847-5G
76650-552EA 60.48 USD
76650-552 76650-554 76650-556 77617-718
tert-Butyl 4-(2-aminopyridin-4-yl)piperazine-1-carboxylate 95%
tert-Butyl 4-(2-aminopyridin-4-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₄O₂
MW: 278.3501 g/mol
MDL Number: MFCD11849292
CAS Number: 571189-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR