1-(2-(4-((4-Carbamoylpiperidin-1-yl)methyl)-N-methylbenzamido)ethyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate 98+%

Supplier: Ambeed
Warning

Synonyms: Revefenacin

A642503-10MG A642503-1MG A642503-25MG A642503-5MG
76650-540EA 39.26 USD
76650-540 76650-542 76650-544 76650-546
1-(2-(4-((4-Carbamoylpiperidin-1-yl)methyl)-N-methylbenzamido)ethyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate 98+%
1-(2-(4-((4-Carbamoylpiperidin-1-yl)methyl)-N-methylbenzamido)ethyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₃₅H₄₃N₅O₄
MW: 597.75 g/mol
MDL Number: MFCD28386316
CAS Number: 864750-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR