[1,1':4',1''-Terphenyl]-4,4''-dicarboxaldehyde 98%

Supplier: Ambeed
Warning

A641759-100MG A641759-5G A641759-1G A641759-250MG
76650-530EA 10 USD
76650-530 76650-532 77749-317 77749-318
[1,1':4',1''-Terphenyl]-4,4''-dicarboxaldehyde 98%
[1,1':4',1''-Terphenyl]-4,4''-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₂
MDL Number: MFCD31560630
CAS Number: 62940-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR