Polyethylene glycol monomethyl ether, M.W. ∼2000

Supplier: Ambeed
Warning

A641450-1000G A641450-100G A641450-10G A641450-25G A641450-500G
76650-518EA 62 USD
76650-518 76650-520 76650-522 76650-524 76650-526
Polyethylene glycol monomethyl ether, M.W. ∼2000
Polyethylene glycol monomethyl ether

Precaución: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR