2-(2-Isopropoxy-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

A641133-1G A641133-250MG A641133-5G
76650-512EA 209 USD
76650-512 76650-514 76650-516
2-(2-Isopropoxy-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2-Isopropoxy-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₂BF₃O₃
MW: 330.1503 g/mol
MDL Number: MFCD28806027
CAS Number: 2121513-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR