Ethyl-4-hydroxypyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A639596-1G A639596-250MG A639596-5G
76650-508EA 37 USD
76650-508 76650-510 77617-696
Ethyl-4-hydroxypyrimidine-5-carboxylate 95%
Ethyl-4-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD00235034
CAS Number: 4786-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR