Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lithium PTMB phosphinate, LAP, Phenyl(2,4,6-trimethylbenzoyl)phosphinic acid lithium salt, Lithium Phenyl(2,4,6-trimethylbenzoyl)phosphinate
A636763-1G
A636763-250MG
A636763-25G
A636763-5G
A636763-100G
76650-492EA
20
USD
76650-492
76650-494
76650-496
76650-498
77783-551
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 97%
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP)
Precaución: for Research Use Only
Formula:
C₁₆H₁₆LiO₃P |
MDL Number:
MFCD29472537 CAS Number: 85073-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...