(S)-2-(Fmoc-amino)heptanoic acid 98%

Supplier: Ambeed
Warning

A636352-10G A636352-1G A636352-250MG A636352-25G A636352-5G
76650-482EA 405.27 USD
76650-482 76650-484 76650-486 76650-488 76650-490
(S)-2-(Fmoc-amino)heptanoic acid 98%
(S)-2-(Fmoc-amino)heptanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
MW: 367.4382 g/mol
MDL Number: MFCD08275892
CAS Number: 1197020-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR