2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxy-3-chloro-5-fluorobenzeneboronic acid pinacol ester

A636322-1G A636322-250MG A636322-5G
76650-476EA 161.26 USD
76650-476 76650-478 76650-480
2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₉H₂₁BClFO₃
MW: 362.6307 g/mol
MDL Number: MFCD30529839
CAS Number: 2121515-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR