4-Fluoro-2-methyl-3-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A635487-100MG
A635487-250MG
A635487-50MG
A635487-5G
A635487-1G
76650-468EA
57
USD
76650-468
76650-470
76650-472
76650-474
77308-030
4-Fluoro-2-methyl-3-nitrobenzoic acid 98%
4-Fluoro-2-methyl-3-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆FNO₄ |
MDL Number:
MFCD28797610 CAS Number: 1079991-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...