2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A635422-100MG A635422-1G A635422-250MG A635422-5G
76650-460EA 34 USD
76650-460 76650-462 76650-464 76650-466
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)aniline 98%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₃NO₂
MW: 287.0858 g/mol
MDL Number: MFCD16996232
CAS Number: 1196972-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR